Merck
  • The role of Ets2 transcription factor in the induction of microRNA-155 (miR-155) by lipopolysaccharide and its targeting by interleukin-10.
The role of Ets2 transcription factor in the induction of microRNA-155 (miR-155) by lipopolysaccharide and its targeting by interleukin-10.
The Journal of biological chemistry (2013-12-24)
Susan R Quinn, Niamh E Mangan, Brian E Caffrey, Michael P Gantier, Bryan R G Williams, Paul J Hertzog, Claire E McCoy, Luke A J O'Neill
ABSTRAKT
MicroRNA-155 (miR-155) is highly expressed in many cancers such as B cell lymphomas and myeloid leukemia and inflammatory disorders such as rheumatoid arthritis, atopic dermatitis, and multiple sclerosis. The role of miR-155 as both a promoter of inflammation and an oncogenic agent provides a clear need for miR-155 itself to be stringently regulated. We therefore investigated the transcriptional regulation of miR-155 in response to the respective pro- and anti-inflammatory mediators LPS and IL-10. Bioinformatic analysis revealed Ets binding sites on the miR-155 promoter, and we found that Ets2 is critical for miR-155 induction by LPS. Truncation and mutational analysis of the miR-155 promoter confirmed the role of the Ets2 binding site proximal to the transcription start site for LPS responsiveness. We observed increased binding of Ets2 to the miR-155 promoter and Ets2 deficient mice displayed decreased induction of miR-155 in response to LPS. IL-10 inhibited the induction of Ets2 mRNA and protein by LPS, thereby decreasing Ets2 function on the pri-155 promoter. We have thus identified Ets2 as a key novel regulator in both the positive and negative control of miR-155 in the inflammatory response.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
STAT3 Inhibitor V, Stattic, STAT3 Inhibitor V, Stattic, CAS 19983-44-9, is a cell-permeable inhibitor of STAT3 cellular function that targets the STAT3-SH2 domain and prevents its association with upstream Kinases.
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution