Merck
Loss of striatal type 1 cannabinoid receptors is a key pathogenic factor in Huntington's disease.
Brain : a journal of neurology (2010-10-12)
Cristina Blázquez, Anna Chiarlone, Onintza Sagredo, Tania Aguado, M Ruth Pazos, Eva Resel, Javier Palazuelos, Boris Julien, María Salazar, Christine Börner, Cristina Benito, Carolina Carrasco, María Diez-Zaera, Paola Paoletti, Miguel Díaz-Hernández, Carolina Ruiz, Michael Sendtner, José J Lucas, Justo G de Yébenes, Giovanni Marsicano, Krisztina Monory, Beat Lutz, Julián Romero, Jordi Alberch, Silvia Ginés, Jürgen Kraus, Javier Fernández-Ruiz, Ismael Galve-Roperh, Manuel Guzmán
ABSTRAKT
Endocannabinoids act as neuromodulatory and neuroprotective cues by engaging type 1 cannabinoid receptors. These receptors are highly abundant in the basal ganglia and play a pivotal role in the control of motor behaviour. An early downregulation of type 1 cannabinoid receptors has been documented in the basal ganglia of patients with Huntington's disease and animal models. However, the pathophysiological impact of this loss of receptors in Huntington's disease is as yet unknown. Here, we generated a double-mutant mouse model that expresses human mutant huntingtin exon 1 in a type 1 cannabinoid receptor-null background, and found that receptor deletion aggravates the symptoms, neuropathology and molecular pathology of the disease. Moreover, pharmacological administration of the cannabinoid Δ(9)-tetrahydrocannabinol to mice expressing human mutant huntingtin exon 1 exerted a therapeutic effect and ameliorated those parameters. Experiments conducted in striatal cells show that the mutant huntingtin-dependent downregulation of the receptors involves the control of the type 1 cannabinoid receptor gene promoter by repressor element 1 silencing transcription factor and sensitizes cells to excitotoxic damage. We also provide in vitro and in vivo evidence that supports type 1 cannabinoid receptor control of striatal brain-derived neurotrophic factor expression and the decrease in brain-derived neurotrophic factor levels concomitant with type 1 cannabinoid receptor loss, which may contribute significantly to striatal damage in Huntington's disease. Altogether, these results support the notion that downregulation of type 1 cannabinoid receptors is a key pathogenic event in Huntington's disease, and suggest that activation of these receptors in patients with Huntington's disease may attenuate disease progression.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-GAD67 Antibody, clone 1G10.2, clone 1G10.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse