Merck
Atg5 regulates late endosome and lysosome biogenesis.
Science China. Life sciences (2013-12-27)
Junya Peng, Ran Zhang, Yitong Cui, Haodong Liu, Xiaoxin Zhao, Lei Huang, Mingxu Hu, Xiaoxi Yuan, Benyu Ma, Xiaowei Ma, Ueno Takashi, Komatsu Masaaki, Xingjie Liang, Li Yu
ABSTRAKT
Autophagy is an evolutionarily conserved lysosome-based degradation process. Atg5 plays a very important role in autophagosome formation. Here we show that Atg5 is required for biogenesis of late endosomes and lysosomes in an autophagy-independent manner. In Atg5 (-/-) cells, but not in other essential autophagy genes defecting cells, recycling and retrieval of late endosomal components from hybrid organelles are impaired, causing persistent hybrid organelles and defective formation of late endosomes and lysosomes. Defective retrieval of late endosomal components from hybrid organelles resulting from impaired recruitment of a component of V1-ATPase to acidic organelles blocks the pH-dependent retrieval of late endosomal components from hybrid organelles. Lowering the intracellular pH restores late endosome/lysosome biogenesis in Atg5 (-/-) cells. Our data demonstrate an unexpected role of Atg5 and shed new light on late endosome and lysosome biogenesis.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-LAMP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Lysosome Isolation Kit, sufficient for 25 g (tissue), sufficient for 20 mL (packed cells), enrichment of lysosomes from tissues and packed cells
Sigma-Aldrich
Anti-LAMP2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 60,000-76,000
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)