Merck
  • Reactivity of antibodies from patients with acute and chronic paracoccidioidomycosis to a high molecular mass antigen from Paracoccidioides brasiliensis.
Reactivity of antibodies from patients with acute and chronic paracoccidioidomycosis to a high molecular mass antigen from Paracoccidioides brasiliensis.
Journal of clinical laboratory analysis (2005-09-20)
A S Marquez, A P Vicentini, M A Ono, M A E Watanabe, Z P de Camargo, E N Itano
ABSTRAKT
Yeast forms of Paracoccidioides brasiliensis produce polydispersed high molecular mass (h-MM) antigens. We investigated the antibodies to an h-MM antigen from P. brasiliensis by immunoblotting and ELISA in sera from paracoccidioidomycosis (PCM) patients. IgG from the sera of chronic PCM patients was able to recognize the h-MM antigen at a higher frequency in the cell-free antigen (CFA) (8/13) than in the somatic antigen (SA) (2/13), as assessed by immunoblotting. The CFA was fractionated by Sephadex G-200 chromatography, and fraction 17 (F17) with the h-MM antigen of approximately 366 kDa was used in ELISA to analyze specific levels of IgG and IgE. Patients with the chronic form showed significantly higher levels of IgG (P<0.05) but not IgE (P>0.05) to F17 by ELISA, compared to patients with the acute form or to healthy donors. In conclusion, CFA is better than SA as a source of the P. brasiliensis h-MM antigen. This study reveals a new characteristic to differentiate between the acute and chronic forms of PCM, by demonstrating a higher level of seric IgG to h-MM antigen in chronic compared to acute PCM patients.