Merck
HIF-2α phosphorylation by CK1δ promotes erythropoietin secretion in liver cancer cells under hypoxia.
Journal of cell science (2016-10-01)
Evanthia Pangou, Christina Befani, Ilias Mylonis, Martina Samiotaki, George Panayotou, George Simos, Panagiotis Liakos
ABSTRAKT
Hypoxia inducible factor 2 (HIF-2) is a transcriptional activator implicated in the cellular response to hypoxia. Regulation of its inducible subunit, HIF-2α (also known as EPAS1), involves post-translational modifications. Here, we demonstrate that casein kinase 1δ (CK1δ; also known as CSNK1D) phosphorylates HIF-2α at Ser383 and Thr528 in vitro We found that disruption of these phosphorylation sites, and silencing or chemical inhibition of CK1δ, reduced the expression of HIF-2 target genes and the secretion of erythropoietin (EPO) in two hepatic cancer cell lines, Huh7 and HepG2, without affecting the levels of HIF-2α protein expression. Furthermore, when CK1δ-dependent phosphorylation of HIF-2α was inhibited, we observed substantial cytoplasmic mislocalization of HIF-2α, which was reversed upon the addition of the nuclear protein export inhibitor leptomycin B. Taken together, these data suggest that CK1δ enhances EPO secretion from liver cancer cells under hypoxia by modifying HIF-2α and promoting its nuclear accumulation. This modification represents a new mechanism of HIF-2 regulation that might allow HIF isoforms to undertake differing functions.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M5 antibody produced in mouse, clone M5, purified immunoglobulin, buffered aqueous solution