Merck
Metabolic Engineering of Escherichia coli for the Production of 3-Hydroxypropionic Acid and Malonic Acid through β-Alanine Route.
ACS synthetic biology (2016-03-02)
Chan Woo Song, Je Woong Kim, In Jin Cho, Sang Yup Lee
ABSTRAKT
Escherichia coli was metabolically engineered to produce industrially important platform chemicals, 3-hydroxypropionic acid (3-HP) and malonic acid (MA), through the β-alanine (BA) route. First, various combinations of downstream enzymes were screened and BA pyruvate transaminase (encoded by pa0132) from P. aeruginosa was selected to generate malonic semialdehyde (MSA) from BA. This platform strain was engineered by introducing E. coli MSA reductase (encoded by ydfG) to reduce MSA to 3-HP. Replacement of native promoter of the sdhC gene with the strong trc promoter in the genome increased 3-HP production to 3.69 g/L in flask culture. Introduction of E. coli semialdehyde dehydrogenase (encoded by yneI) into the platform strain resulted in the production of MA, and additional deletion of the ydfG gene increased MA production to 0.450 g/L in flask culture. Fed-batch cultures of final engineered strains resulted in the production of 31.1 g/L 3-HP or 3.60 g/L MA from glucose.