Merck
The Axon Protective Effects of Syringic Acid on Ischemia/Reperfusion Injury in a Rat Sciatic Nerve Model.
Turkish neurosurgery (2015-07-06)
Mehmet Tokmak, Muserref Hilal Sehitoglu, Yasemin Yuksel, Mustafa Guven, Tarik Akman, Adem Bozkurt Aras, Umut Yaka, Cengiz Gomleksiz, Serdar Baki Albayrak, Murat Cosar
ABSTRAKT
In the relevant literature, there is no experimental study that investigated the axon protective effects of syringic acid- a polyphenol compound- with an anti-oxidant capacity on ischemia/reperfusion injury. The rats were randomly divided into four groups: Control group (no medication or surgical procedure), Sham group, Syringic acid group, and Methyprednisolone (MP) Group. Ischemia was achieved by abdominal aorta clamping and all animals were sacrificed 24 hours after ischemia. Harvested sciatic nerve segments were investigated histopathologically and for tissue biochemistry. Ischemic fiber degeneration scores were found significantly lower in syringic acid and MP groups than sham group. Additionally, apoptosis-related cysteine peptidase caspase-3 immunostaining scores were lower in syringic acid and MP groups. Biochemically, superoxide dismutase and nuclear respiratory factor 1 values were significantly higher in syringic acid group compared to those of control and sham groups while malondialdehyde levels were significantly lower in the syringic acid group. Syringic acid reduces oxidative stress and axonal degeneration in rat sciatic nerve after ischemia/reperfusion injury. Therefore, syringic acid may play a role in the treatment of peripheral nerve injuries due to ischemia/reperfusion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Caspase 3 Antibody, active (cleaved) form, Chemicon®, from rabbit
Sigma-Aldrich
Zestaw testowy SOD, sufficient for 500 tests
Sigma-Aldrich
BrdU (5-bromo-2′-deoksyurydyna), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.