- Intact mitochondrial Ca
Intact mitochondrial Ca
Free radical biology & medicine (2016-12-08)
Suphachai Charoensin, Emrah Eroglu, Marissa Opelt, Helmut Bischof, Corina T Madreiter-Sokolowski, Andrijana Kirsch, Maria R Depaoli, Saša Frank, Astrid Schrammel, Bernd Mayer, Markus Waldeck-Weiermair, Wolfgang F Graier, Roland Malli
PMID27923677
ABSTRAKT
Mitochondrial Ca