- Synthesis, Acaricidal Activity, and Structure-Activity Relationships of Pyrazolyl Acrylonitrile Derivatives.
Synthesis, Acaricidal Activity, and Structure-Activity Relationships of Pyrazolyl Acrylonitrile Derivatives.
Journal of agricultural and food chemistry (2016-12-16)
Haibo Yu, Yan Cheng, Man Xu, Yuquan Song, Yanmei Luo, Bin Li
PMID27976890
ABSTRAKT
A series of novel pyrazolyl acrylonitrile derivatives was designed, targeting Tetranychus cinnabarinus, and synthesized. Their structures were identified by combination of