Merck
Synthesis of thiolated polysaccharides for formation of polyelectrolyte multilayers with improved cellular adhesion.
Carbohydrate polymers (2016-12-19)
Pegah Esmaeilzadeh, Alexander Köwitsch, Frank Heyroth, Georg Schmidt, Steffen Fischer, Katja Richter, Thomas Groth
ABSTRAKT
Intrinsic cross-linking is not only useful for increasing stability, but also for tailoring mechanical properties of polyelectrolyte multilayers (PEM) on implants and tissue engineering scaffolds. Here, a novel route for synthesizing thiolated chitosan (t-Chi) based on the application of 3,3'-dithiodipropionic acid was applied, while thiolated chondroitin sulfate (t-CS) was conjugated by 3,3'-dithiobis (propanoic hydrazide). Both products were subsequently reduced to obtain the free thiols. The thiol content, structural changes and degree of substitution were studied by UV-vis, FTIR, Raman and
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
11-Mercaptoundecanoic acid, 95%
Sigma-Aldrich
3,3′-Dithiodipropionic acid, 95%
Sigma-Aldrich
2-Vinylpyridine, 97%