Merck
Recurrent bladder carcinoma: clinical and prognostic role of 18 F-FDG PET/CT.
European journal of nuclear medicine and molecular imaging (2016-08-28)
Pierpaolo Alongi, Federico Caobelli, Roberta Gentile, Alessandro Stefano, Giorgio Russo, Domenico Albano, Sergio Baldari, Maria Carla Gilardi, Massimo Midiri
ABSTRAKT
A small number of studies evaluated the detection rate of lesions from bladder carcinoma (BC) of 18 F-FDG PET/CT in the restaging process. However, the prognostic role of FDG PET/CT still remains unclear. The aim of the present study was to evaluate the accuracy, the effect upon treatment decision, and the prognostic value of FDG PET/CT in patients with suspected recurrent BC. Forty-one patients affected by BC underwent FDG PET/CT for restaging purpose. The diagnostic accuracy of visually interpreted FDG PET/CT was assessed compared to histology (n = 8), other diagnostic imaging modalities (contrast-enhanced CT in 38/41 patients and MRI in 15/41) and clinical follow-up (n = 41). Semiquantitative PET values (SUVmax, SUVmean, SUL, MTV, TLG) were calculated using a graph-based method. Progression-free survival (PFS) and overall survival (OS) were assessed by using Kaplan-Meier curves. The risk of progression (hazard ratio, HR) was computed by Cox regression analysis by considering all the available variables. PET was considered positive in 21 of 41 patients. Of these, recurrent BC was confirmed in 20 (95 %). Sensitivity, specificity, positive predictive value, negative predictive value, and accuracy of FDG PET/CT were 87 %, 94 %, 95 %, 85 %, 90 %. AUC was 0.9 (95 %IC 0.8-1). Bayesian positive and negative likelihood ratios were 14.5 and 0.13, respectively. FDG PET/CT findings modified the therapeutic approach in 16 patients (modified therapy in 10 PET-positive patients, watch-and-wait in six PET-negative patients). PFS was significantly longer in patients with negative scan vs. those with pathological findings (85 % vs. 24 %, p < 0.05; HR = 12.4; p = 0.001). Moreover, an unremarkable study was associated with a longer OS (88 % vs. 47 % after 2 years and 87 % vs. 25 % after 3 years, respectively, p < 0.05). Standardized uptake value (SUV)max > 6 and total lesion glycolysis (TLG) > 8.5 were recognized as the most accurate thresholds to predict PFS (2-year PFS 62 % for SUVmax < 6 vs. 15 % for SUVmax > 6, p = 0.018; 2-year PFS 66 % for TLG < 8.5 vs. 18 % for TLG > 8.5, p = 0.09). A very good diagnostic performance for FDG PET/CT was confirmed in patients with suspected recurrent BC. FDG PET/CT allowed for a change in treatment decision in about 40 % of cases and showed an important prognostic value in assessing PFS and OS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Fluorescein di(β-D-galactopyranoside)