Merck
  • Descriptive and multivariate analyses of four Tunisian wastewater treatment plants: A comparison between different treatment processes and their efficiency improvement.
Descriptive and multivariate analyses of four Tunisian wastewater treatment plants: A comparison between different treatment processes and their efficiency improvement.
Journal of environmental management (2016-11-25)
Ines Boujelben, Youssef Samet, Mouna Messaoud, Mohamed Ben Makhlouf, Sami Maalej
ABSTRAKT
This study was undertaken to evaluate the performance of four wastewater treatment plants/processes over a 4 year period. The wastewater flow evolution, energy consumption, and quality indicator parameters (BOD