Merck
  • Synthesis and pharmacologic evaluation of 2-endo-amino-3-exo-isopropylbicyclo[2.2.1]heptane: a potent imidazoline1 receptor specific agent.
Synthesis and pharmacologic evaluation of 2-endo-amino-3-exo-isopropylbicyclo[2.2.1]heptane: a potent imidazoline1 receptor specific agent.
Journal of medicinal chemistry (1996-03-15)
S A Munk, R K Lai, J E Burke, P N Arasasingham, A B Kharlamb, C A Manlapaz, E U Padillo, M K Wijono, D W Hasson, L A Wheeler, M E Garst