Merck
The Atypical Dual Specificity Phosphatase hYVH1 Associates with Multiple Ribonucleoprotein Particles.
The Journal of biological chemistry (2016-11-20)
Qiudi Geng, Besa Xhabija, Colleen Knuckle, Christopher A Bonham, Panayiotis O Vacratsis
ABSTRAKT
Human YVH1 (hYVH1), also known as dual specificity phosphatase 12 (DUSP12), is a poorly characterized atypical dual specificity phosphatase widely conserved throughout evolution. Recent findings have demonstrated that hYVH1 expression affects cellular DNA content and is a novel cell survival phosphatase preventing both thermal and oxidative stress-induced cell death, whereas studies in yeast have established YVH1 as a novel 60S ribosome biogenesis factor. In this study, we have isolated novel hYVH1-associating proteins from human U2OS osteosarcoma cells using affinity chromatography coupled to mass spectrometry employing ion mobility separation. Numerous ribosomal proteins were identified, confirming the work done in yeast. Furthermore, proteins known to be present on additional RNP particles were identified, including Y box-binding protein 1 (YB-1) and fragile X mental retardation protein, proteins that function in translational repression and stress granule regulation. Follow-up studies demonstrated that hYVH1 co-localizes with YB-1 and fragile X mental retardation protein on stress granules in response to arsenic treatment. Interestingly, hYVH1-positive stress granules were significantly smaller, whereas knocking down hYVH1 expression attenuated stress granule breakdown during recovery from arsenite stress, indicating a possible role for hYVH1 in stress granule disassembly. These results propagate a role for dual specificity phosphatases at RNP particles and suggest that hYVH1 may affect a variety of fundamental cellular processes by regulating messenger ribonucleoprotein (mRNP) dynamics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Ribosomal Protein L26 (N-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)