Merck
  • Diglycolamide-functionalized poly(propylene imine) diaminobutane dendrimers for sequestration of trivalent f-elements: synthesis, extraction and complexation.
Diglycolamide-functionalized poly(propylene imine) diaminobutane dendrimers for sequestration of trivalent f-elements: synthesis, extraction and complexation.
Dalton transactions (Cambridge, England : 2003) (2016-12-15)
Andrea Leoncini, Seraj A Ansari, Prasanta K Mohapatra, Arijit Sengupta, Jurriaan Huskens, Willem Verboom
ABSTRAKT
Three diglycolamide-functionalized poly(propylene imine) diaminobutane dendrimers, viz., zero generation (LI), first generation (LII), and second generation (LIII), were synthesized and evaluated for their complexation ability towards trivalent actinides and lanthanides. The distribution coefficient (D) of Am
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis