Merck
A characteristic biosignature for discrimination of gastric cancer from healthy population by high throughput GC-MS analysis.
Oncotarget (2016-09-03)
Yinan Chen, Jun Zhang, Lei Guo, Lei Liu, Jingran Wen, Lu Xu, Min Yan, Zuofeng Li, Xiaoyan Zhang, Peng Nan, Jinling Jiang, Jun Ji, Jianian Zhang, Wei Cai, Huisheng Zhuang, Yan Wang, Zhenggang Zhu, Yingyan Yu
ABSTRAKT
Early diagnosis of gastric cancer is crucial to improve patient' outcome. A good biomarker will function in early diagnosis for gastric cancer. In order to find practical and cost-effective biomarkers, we used gas chromatography combined mass spectrometer (GC-MS) to profile urinary metabolites on 293 urine samples. Ninety-four samples are taken as training set, others for validating study. Orthogonal partial least squares discriminant analysis (OPLS-DA), significance analysis of microarray (SAM) and Mann-Whitney U test are used for data analysis. The diagnostic value of urinary metabolites was evaluated by ROC curve. As results, Seventeen metabolites are significantly different between patients and healthy controls in training set. Among them, 14 metabolites show diagnostic value better than classic blood biomarkers by quantitative assay on validation set. Ten of them are amino acids and four are organic metabolites. Importantly, proline, p-cresol and 4-hydroxybenzoic acid disclose outcome-prediction value by means of survival analysis. Therefore, the examination of urinary metabolites is a promising noninvasive strategy for gastric cancer screening.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethyl 2-methylacetoacetate, 90%