Merck
Gradient-Hierarchic-Aligned Porosity SiOC Ceramics.
Scientific reports (2017-01-21)
Cekdar Vakifahmetoglu, Damla Zeydanli, Murilo Daniel de Mello Innocentini, Fernanda Dos Santos Ribeiro, Paulo Renato Orlandi Lasso, Gian Domenico Soraru
ABSTRAKT
This work describes a simple technique to produce porous ceramics with aligned porosity having very high permeability and specific surface area. SiOC-based compositions were processed from blends of three types of preceramic polymer and a catalyst, followed by curing and pyrolysis. The heating applied from the bottom of molds promoted the nucleation, expansion and rising of gas bubbles, and the creation of a ceramic matrix with axially oriented channels interconnected by small round pores. The samples were analyzed by SEM, tomography, BET, water immersion porosimetry and permeation to gas flow. The resulting bodies presented levels of open porosity (69.9-83.4%), average channel diameter (0.59-1.25 mm) and permeability (0.56-3.83 × 10
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)