Merck
  • Comparison of lidocaine/tetracaine cream and lidocaine/prilocaine cream for local anaesthesia during laser treatment of acne keloidalis nuchae and tattoo removal: results of two randomized controlled trials.
Comparison of lidocaine/tetracaine cream and lidocaine/prilocaine cream for local anaesthesia during laser treatment of acne keloidalis nuchae and tattoo removal: results of two randomized controlled trials.
The British journal of dermatology (2016-07-06)
K Greveling, E P Prens, N Ten Bosch, M B van Doorn
ABSTRAKT
Pain is a common adverse effect of dermatological laser procedures. Currently, no standard topical anaesthetic cream exists for deeper dermal laser procedures. To compare the efficacy of lidocaine/tetracaine cream and lidocaine/prilocaine cream in reducing self-reported pain during deeper dermal laser treatment of acne keloidalis nuchae (AKN) and tattoos. We conducted two randomized, double-blind, controlled clinical trials with intrapatient, split-lesion designs: study A included patients with AKN (n = 15); study B included patients with black tattoos (n = 15). The primary end point was the patients' self-reported pain on a 10-cm visual analogue scale (VAS). Secondary objectives were the percentage of patients with adequate pain relief, willingness to pay €25 for the cream that provided the best pain relief and safety of the creams. In both studies, VAS scores were lower for lidocaine/prilocaine cream, with a mean VAS difference in study A of 1·9 [95% confidence interval (CI) 1·0-2·8] and in study B of 0·6 (95% CI -0·7 to 1·9). In study A, adequate pain relief was achieved in 13% (n = 2) with lidocaine/tetracaine cream vs. 73% (n = 11) with lidocaine/prilocaine cream (P = 0·004), and in study B in 53% (n = 8) vs. 80% (n = 12), respectively (P = 0·289). In study A, 47% (n = 7) were willing to pay an additional €25 vs. 73% (n = 11) in study B. No serious adverse events occurred. Lidocaine/prilocaine cream under plastic occlusion is the preferred topical anaesthetic during painful laser procedures targeting dermal chromophores.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Yttrium aluminum oxide, nanopowder, <150 nm particle size (TEM), 99% trace metals basis