Merck
Phosphonopeptides, a new class of synthetic antibacterial agents.
Nature (1978-03-02)
J G Allen, F R Atherton, M J Hall, C H Hassall, S W Holmes, R W Lambert, L J Nisbet, P S Ringrose
PMID628432