Merck
  • A 4-week study assessing the pharmacokinetics, pharmacodynamics, safety, and tolerability of the glucagon receptor antagonist PF-06291874 administered as monotherapy in subjects with type 2 diabetes mellitus.
A 4-week study assessing the pharmacokinetics, pharmacodynamics, safety, and tolerability of the glucagon receptor antagonist PF-06291874 administered as monotherapy in subjects with type 2 diabetes mellitus.
Diabetes research and clinical practice (2017-02-27)
Arthur Bergman, Beesan Tan, Veena R Somayaji, Roberto A Calle, David J Kazierad
ABSTRAKT
The glucagon receptor antagonist PF-06291874 has demonstrated robust glucose reductions in subjects with type 2 diabetes mellitus (T2DM) on background metformin. This study assessed the pharmacokinetics, pharmacodynamics, safety, and tolerability of PF-06291874 administered as monotherapy in subjects with T2DM. After a ≥4-week antidiabetic therapy washout period, 172 subjects were randomized to placebo or PF-06291874 15, 35, 75, or 150mg once daily for 28days. Mean daily glucose (MDG), fasting plasma glucose (FPG), and predefined safety endpoints were assessed at baseline and day 28. Dose-dependent reductions (placebo-adjusted) from baseline in MDG ranged from 40.3 to 68.8mg/dL and in FPG from 27.1 to 57.2mg/dL after 28days of dosing with PF-06291874. There were no significant changes in low-density lipoprotein cholesterol at doses ≤75mg relative to placebo. Small, dose-dependent increases in alanine aminotransferase and aspartate aminotransferase were observed; however, the incidence of these values >3×upper limit of normal was similar across doses. PF-06291874 exposures were consistent with previous studies and PF-06291874 was well tolerated, with minimal incidence of hypoglycemia. PF-06291874 as monotherapy was well tolerated and produced robust reductions in plasma glucose following 4weeks of dosing in subjects with T2DM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
PF-06291874, ≥98% (HPLC)