Merck
Relationships between the structures of flavanone derivatives and their effects in enhancing early growth response-1 gene expression.
Bioorganic & medicinal chemistry letters (2009-03-25)
Sunhee Lee, Yoonkyung Woo, Soon Young Shin, Young Han Lee, Yoongho Lim
ABSTRAKT
To identify the structural requirements that are pivotal in enhancing Early growth response-1 (Egr-1) expression, the quantitative relationships between the structural properties of flavanone derivatives and their increments of Egr-1 expression were elucidated using comparative molecular field analysis and comparative molecular similarity indices analysis.