Merck
  • Analysis of amino acid and monoamine neurotransmitters and their metabolites in rat urine of Alzheimer's disease using in situ ultrasound-assisted derivatization dispersive liquid-liquid microextraction with UHPLC-MS/MS.
Analysis of amino acid and monoamine neurotransmitters and their metabolites in rat urine of Alzheimer's disease using in situ ultrasound-assisted derivatization dispersive liquid-liquid microextraction with UHPLC-MS/MS.
Journal of pharmaceutical and biomedical analysis (2017-01-01)
Xian-En Zhao, Yongrui He, Meng Li, Guang Chen, Na Wei, Xiao Wang, Jing Sun, Shuyun Zhu, Jinmao You
ABSTRAKT
Neurotransmitters (NTs) may play an important role in neurodegenerative disorders such as Alzheimer's disease (AD). In order to investigate the potential links, a new simple, fast, accurate and sensitive analytical method, based on in situ ultrasound-assisted derivatization dispersive liquid-liquid microextraction (in situ UA-DDLLME) coupled with ultra high-performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS), has been developed and validated. The quantitation of amino acid neurotransmitters (AANTs) and monoamine neurotransmitters (MANTs) in urine of AD rats were performed in this work. The in situ UA-DDLLME procedure involved the rapid injection of the mixture of low toxic 4-bromoanisole (extractant) and acetonitrile (dispersant), which containing the new designed and synthesized 4'-carbonyl chloride rosamine (CCR) as derivatization reagent, into the aqueous phase of real sample and buffer. Under the selected conditions, the derivatization and microextraction of analytes were simultaneously completed within 1min. Good linearity for each analyte (R>0.992) was observed with low limit of detections (LODs, S/N>3). Moreover, the proposed method was compared with direct detection or other reported methods, and the results showed that low matrix effects and good recoveries results were obtained in this work. Taken together, in situ UA-DDLLME coupled with UHPLC-MS/MS analysis was demonstrated to be a good method for sensitive, accurate and simultaneous monitoring of AANTs and MANTs. This method would be expected to be highly useful in AD diseases' clinical diagnostics and may have potential value in monitoring the efficacy of treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%
Sigma-Aldrich
DL-3,4-Dihydroxyphenyl glycol