- Grafting of aniline derivatives onto chitosan and their applications for removal of reactive dyes from industrial effluents.
Grafting of aniline derivatives onto chitosan and their applications for removal of reactive dyes from industrial effluents.
International journal of biological macromolecules (2016-11-26)
Mojtaba Abbasian, Mehdi Jaymand, Pouneh Niroomand, Amir Farnoudian-Habibi, Saber Ghasemi Karaj-Abad
PMID27884669
ABSTRAKT
A series of chitosan-grafted polyaniline derivatives {chitosan-g-polyaniline (CS-g-PANI), chitosan-g-poly(N-methylaniline) (CS-g-PNMANI), and chitosan-g-poly(N-ethylaniline) (CS-g-PNEANI)} were synthesized by in situ chemical oxidation polymerization method. The synthesized copolymers were analyzed by means of Fourier transform infrared (FTIR), and ultraviolet-visible (UV-vis) spectroscopies, thermogravimetric analysis (TGA), and field emission scanning electron microscopy (FE-SEM). These copolymers were applied as adsorbent for removal of acid red 4 (AR4) and direct red 23 (DR23) from aqueous solutions. The adsorption processes were optimized in terms of pH, adsorbent amount, and dyes concentrations. The maximum adsorption capacities (Q