Merck
Hepatic chemerin mRNA expression is reduced in human nonalcoholic steatohepatitis.
European journal of clinical investigation (2016-11-01)
Rebekka Pohl, Elisabeth M Haberl, Lisa Rein-Fischboeck, Sebastian Zimny, Maximilian Neumann, Charalampos Aslanidis, Doris Schacherer, Sabrina Krautbauer, Kristina Eisinger, Thomas S Weiss, Christa Buechler
ABSTRAKT
Chemerin is associated with insulin resistance and is expressed in the liver. Nonalcoholic fatty liver disease (NAFLD) is related to impaired insulin sensitivity, but studies evaluating hepatic and serum chemerin in NAFLD resulted in discordant data. Chemerin mRNA was determined in the liver tissue obtained from 33 controls and 76 NAFLD patients. Chemerin serum levels were measured in a different cohort of patients with ultrasound-diagnosed NAFLD and the respective controls. Hepatic stellate cells and hepatocytes were exposed to selected metabolites and nuclear receptor agonists to study the regulation of chemerin. Effect of recombinant chemerin on hepatocyte released proteins was analysed. Hepatic chemerin expression was not related to BMI, gender, type 2 diabetes and hypertension. Chemerin mRNA did not correlate with steatosis and was negatively associated with inflammation, fibrosis and nonalcoholic steatohepatitis (NASH) score. Patients with NASH had lower chemerin mRNA compared to those with borderline NASH and controls. Factors with a role in NASH mostly did not regulate chemerin in the liver cells. Of note, liver X receptor agonist reduced chemerin protein. Serum chemerin was not changed in NAFLD. Levels positively correlated with age, waist-to-hip ratio, systolic blood pressure, serum FGF21 and lipocalin 2, and negatively with transferrin saturation. Chemerin induced FGF21 in supernatants of primary human hepatocytes. Hepcidin, a major regulator of iron homoeostasis and lipocalin 2, were not regulated by chemerin. Chemerin mRNA is reduced in the liver of NASH patients, and liver X receptor seems to have a role herein.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
GW4064, ≥97% (HPLC)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)