Merck
Autophagy in Dendritic Cells and B Cells Is Critical for the Inflammatory State of TLR7-Mediated Autoimmunity.
Journal of immunology (Baltimore, Md. : 1950) (2016-12-30)
Chi G Weindel, Lauren J Richey, Abhiruchi J Mehta, Mansi Shah, Brigitte T Huber
ABSTRAKT
Individuals suffering from autoimmune disorders possess a hyperactive cellular phenotype where tolerance to self-antigens is lost. Autophagy has been implicated in both the induction and prevention of autoimmunity, and modulators of this cellular recycling process hold high potential for the treatment of autoimmune diseases. In this study, we determine the effects of a loss of autophagy in dendritic cells (DCs), as well as both B cells and DCs, in a TLR7-mediated model of autoimmunity, similar to systemic lupus erythematosus, where both cell types are critical for disease. Although a loss of DC autophagy slowed disease, the combined loss of autophagy in both cell types resulted in a lethal sepsis-like environment, which included tissue inflammation and hyperproduction of inflammasome-associated cytokines. Ablation of B cell signaling reversed this phenotype, indicating that activation of these cells is an essential step in disease induction. Thus, autophagy plays a dichotomous role in this model of disease.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Formalin solution, neutral buffered, 10%, histological tissue fixative
Sigma-Aldrich
Anti-Mouse IgG (γ-chain specific)−FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution