Merck
  • Selective recognition of cis-trans-isomers of platinum drugs and the detection of triplex DNA based on fluorescence reversible model of quantum dots.
Selective recognition of cis-trans-isomers of platinum drugs and the detection of triplex DNA based on fluorescence reversible model of quantum dots.
Journal of pharmaceutical and biomedical analysis (2016-11-28)
Xiaoling Xu, Fang Gao, Xincai Xiao, Yan Hu, Chaozhen Zhu, Dan Zhao
ABSTRAKT
The identification of spatial structures of drugs and the researches on their interaction mechanism with DNA are always attractive to the researchers. However, their realization is lack of simple and fast method. This paper reports the establishment of multiple-functional detection platform based on the "turn off-on" model of ZnCdSe quantum dots. In this system, ZnCdSe quantum dots work as the fluorescent probe, platinum anti-cancer drugs as the quencher and triplex DNA as the trapping agent. The seemingly similar cisplatin and transplatin exhibited different fluorescent recovery behaviors due to their difference in structure, and thus realized the selective detection of cisplatin and transplatin with the reaction time set at 10min as well as the quantitation of cisplatin over the range of 2.5×10
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)