Merck
Norcocaine and cocaethylene distribution patterns in hair samples from light, moderate, and heavy cocaine users.
Drug testing and analysis (2015-12-02)
Cristiana Gambelunghe, Riccardo Rossi, Kyriaki Aroni, Alessio Gili, Mauro Bacci, Vincenzo Pascali, Nadia Fucci
ABSTRAKT
Even though hair analysis often seems to be the best choice for retrospective monitoring of cocaine intake, differentiating between incorporated cocaine and external contamination is widely debated. In this study we report results obtained in 90 hair samples from addicts. All samples were analyzed for cocaine, benzoylecgonine, norcocaine, cocaethylene, and tropococaine by gas chromatography-mass spectrometry (GC-MS) techniques coupled with direct immersion solid-phase micro-extraction. Cocaine concentrations were stratified into three classes of usage: light (0.5-3 ng/mg), moderate (3.1-10 ng/mg) and heavy (10.1-40 ng/mg). The Substance Abuse and Mental Health Services Administration cut-off criteria for establishing active cocaine use were applied to the results. For all samples criteria were cocaine levels above 0.5 ng/mg (ranging from 1.63 to 39.29 ng/mg, mean 9.49 ng/mg), benzoylecgonine concentrations ≥ 0.05 ng/mg (ranging from 0.19 to 5.77 ng/mg, mean 1.40), and benzoylecgonine to cocaine % ratio ≥5% (from 6.43 to 26.09%). Norcocaine was present in 58.9% of samples (concentration range: 0.22-3.14 ng/mg) and was strongly predictive only of heavy cocaine use (sensitivity 100% for cocaine concentrations above 9.58 ng/mg). Twenty hair samples from moderate and heavy users tested positive for cocaethylene (concentration range: 0.22-1.98 ng/mg, mean 0.73 ng/mg). This study on hair samples with no chance of false positive cases highlights the very limited applications of testing minor cocaine metabolites for definitive proof of active cocaine consumption. © 2015 The Authors. Drug Testing and Analysis Published by John Wiley & Sons, Ltd.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
SPME Overcoated Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, notched pink hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub