Merck
  • Hydrogen sulfide alleviates mercury toxicity by sequestering it in roots or regulating reactive oxygen species productions in rice seedlings.
Hydrogen sulfide alleviates mercury toxicity by sequestering it in roots or regulating reactive oxygen species productions in rice seedlings.
Plant physiology and biochemistry : PPB (2016-12-13)
Zhen Chen, Moshun Chen, Ming Jiang
ABSTRAKT
Soil mercury (Hg) contamination is a major factor that affects agricultural yield and food security. Hydrogen sulfide (H
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%