Merck
  • Sirtuin 1 protects the aging heart from contractile dysfunction mediated through the inhibition of endoplasmic reticulum stress-mediated apoptosis in cardiac-specific Sirtuin 1 knockout mouse model.
Sirtuin 1 protects the aging heart from contractile dysfunction mediated through the inhibition of endoplasmic reticulum stress-mediated apoptosis in cardiac-specific Sirtuin 1 knockout mouse model.
International journal of cardiology (2016-11-23)
Yu-Juei Hsu, Shih-Che Hsu, Chiao-Po Hsu, Yen-Hui Chen, Yung-Lung Chang, Junichi Sadoshima, Shih-Ming Huang, Chien-Sung Tsai, Chih-Yuan Lin
ABSTRAKT
The longevity regulator Sirtuin 1 is an NAD We evaluated cardiac contractile function, endoplasmic reticulum stress, apoptosis, and oxidative stress in 6- and 12month-old cardiac-specific Sirtuin 1 knockout (Sirt1 Compared with 6-month-old Sirt1 The present study demonstrates that the aging heart is characterized by contractile dysfunction associated with increased oxidative stress and endoplasmic reticulum stress and Sirtuin 1 might have the ability to protect the aging hearts from the inhibition of endoplasmic reticulum-mediated apoptosis.