Merck
X-linked ectodermal dysplasia receptor (XEDAR) gene silencing prevents caspase-3-mediated apoptosis in Sjögren's syndrome.
Clinical and experimental medicine (2015-12-15)
Margherita Sisto, Loredana Lorusso, Sabrina Lisi
ABSTRAKT
Despite recent advancements in the knowledge of the etiology and pathogenic mechanisms, treatment of the autoimmune disease Sjögren's syndrome (SS) remains mostly empiric and symptom-based, indicating the need for novel therapeutic approaches. Ectodysplasin-A2 (EDA-A2) is a recently isolated member of the tumor necrosis factor superfamily that binds to X-linked ectodermal dysplasia receptor (XEDAR). In this report, we have analyzed the expression and the biological activity of EDA-A2 in human salivary gland epithelial cells (SGEC) from primary Sjögren's syndrome (pSS) patients. We report that EDA-A2 and its receptor XEDAR are overexpressed in pSS SGEC in comparison with healthy individuals and that the EDA-A2/XEDAR system in these cells is involved in the induction of apoptosis via caspases activation. Collectively, our results suggest that EDA-A2/XEDAR system may be a promising agent for the gene therapy of pSS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human EDA2R