- Nickel-Salen supported paramagnetic nanoparticles for 6-His-target recombinant protein affinity purification.
Nickel-Salen supported paramagnetic nanoparticles for 6-His-target recombinant protein affinity purification.
Journal of chromatography. A (2017-02-22)
Zahra Rashid, Ramin Ghahremanzadeh, Mohammad-Reza Nejadmoghaddam, Mahboobeh Nazari, Mohammad-Reza Shokri, Hossein Naeimi, Amir-Hassan Zarnani
PMID28215401
ABSTRAKT
In this research, a simple, efficient, inexpensive, rapid and high yield method for the purification of 6×histidine-tagged recombinant protein was developed. For this purpose, manganese ferrite magnetic nanoparticles (MNPs) were synthesized through a co-precipitation method and then they were conveniently surface-modified with tetraethyl orthosilicate (TEOS) in order to prevent oxidation and form high density of hydroxyl groups. Next, the salen ligand was prepared from condensation reaction of salicylaldehyde and 3-aminopropyl (trimethoxy) silane (APTMS) in 1:1 molar ratio; followed by complexation with Ni(OAc)