Merck
  • Sodium chlorate, a herbicide and major water disinfectant byproduct, generates reactive oxygen species and induces oxidative damage in human erythrocytes.
Sodium chlorate, a herbicide and major water disinfectant byproduct, generates reactive oxygen species and induces oxidative damage in human erythrocytes.
Environmental science and pollution research international (2016-11-01)
Shaikh Nisar Ali, Mir Kaisar Ahmad, Riaz Mahmood
ABSTRAKT
Sodium chlorate (NaClO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Ouabain, European Pharmacopoeia (EP) Reference Standard