Merck
Mitochondria-penetrating peptides conjugated to desferrioxamine as chelators for mitochondrial labile iron.
PloS one (2017-02-09)
Roxana Y P Alta, Hector A Vitorino, Dibakar Goswami, Cleber W Liria, Simon P Wisnovsky, Shana O Kelley, M Terêsa Machini, Breno P Espósito
ABSTRAKT
Desferrioxamine (DFO) is a bacterial siderophore with a high affinity for iron, but low cell penetration. As part of our ongoing project focused on DFO-conjugates, we synthesized, purified, characterized and studied new mtDFOs (DFO conjugated to the Mitochondria Penetrating Peptides TAT49-57, 1A, SS02 and SS20) using a succinic linker. These new conjugates retained their strong iron binding ability and antioxidant capacity. They were relatively non toxic to A2780 cells (IC50 40-100 μM) and had good mitochondrial localization (Rr +0.45 -+0.68) as observed when labeled with carboxy-tetramethylrhodamine (TAMRA) In general, mtDFO caused only modest levels of mitochondrial DNA (mtDNA) damage. DFO-SS02 retained the antioxidant ability of the parent peptide, shown by the inhibition of mitochondrial superoxide formation. None of the compounds displayed cell cycle arrest or enhanced apoptosis. Taken together, these results indicate that mtDFO could be promising compounds for amelioration of the disease symptoms of iron overload in mitochondria.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Fmoc-Lys(Boc)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.