Merck
A novel fluorescent aptasensor for ultrasensitive detection of microcystin-LR based on single-walled carbon nanotubes and dapoxyl.
Talanta (2017-02-19)
Seyed Mohammad Taghdisi, Noor Mohammad Danesh, Mohammad Ramezani, Narjes Ghows, Seyed Ali Mousavi Shaegh, Khalil Abnous
ABSTRAKT
To assure water safety and protect human health, precise and simple analytical approaches are highly desired to determine low concentrations of microcystin-leucine-arginine (MC-LR), a toxin, in both water and serum samples. Herein, a simple, rapid and accurate aptamer-based fluorescent sensor was used for selective recognition of MC-LR, based on single-walled carbon nanotubes (SWNTs) as immobilizers, dapoxyl as a fluorescent dye, DAP-10 as a specific aptamer for dapoxyl and unmodified MC-LR aptamer (Apt) as a sensing ligand. The sensing method was developed to produce a remarkable fluorescence intensity difference in the absence and presence of MC-LR. Moreover, the Apt was used without any modification. In the absence of MC-LR, the dapoxyl could bind to DAP-10, leading to a strong fluorescence intensity. In the presence of MC-LR, DAP-10 bound to the surface of SWNTs, resulting in a very weak fluorescence intensity. Under optimized conditions, the presented fluorescent analytical approach showed high selectivity toward MC-LR with a limit of detection (LOD) of 138 pM. This new method indicated excellent analytical performance for MC-LR detection in tap water and serum samples with LODs of 135 and 168 pM, respectively.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
N-(6-Chloro-3-pyridylmethyl)-N-cyano-N-methylacetamidine, PESTANAL®, analytical standard