Merck
Discovery of
Proceedings of the National Academy of Sciences of the United States of America (2017-03-23)
Dainan Mao, Leah B Bushin, Kyuho Moon, Yihan Wu, Mohammad R Seyedsayamdost
ABSTRAKT
Bacteria produce a diverse array of secondary metabolites that have been invaluable in the clinic and in research. These metabolites are synthesized by dedicated biosynthetic gene clusters (BGCs), which assemble architecturally complex molecules from simple building blocks. The majority of BGCs in a given bacterium are not expressed under normal laboratory growth conditions, and our understanding of how they are silenced is in its infancy. Here, we have addressed this question in the Gram-negative model bacterium
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
2-Hydroxyquinoline, 98%