Merck
Human Endometrial Stromal Cells Are Highly Permissive To Productive Infection by Zika Virus.
Scientific reports (2017-03-11)
Isabel Pagani, Silvia Ghezzi, Adele Ulisse, Alicia Rubio, Filippo Turrini, Elisabetta Garavaglia, Massimo Candiani, Concetta Castilletti, Giuseppe Ippolito, Guido Poli, Vania Broccoli, Paola Panina-Bordignon, Elisa Vicenzi
ABSTRAKT
Zika virus (ZIKV) is a recently re-emerged flavivirus transmitted to humans by mosquito bites but also from mother to fetus and by sexual intercourse. We here show that primary human endometrial stromal cells (HESC) are highly permissive to ZIKV infection and support its in vitro replication. ZIKV envelope expression was detected in the endoplasmic reticulum whereas double-stranded viral RNA colocalized with vimentin filaments to the perinuclear region. ZIKV productive infection also occurred in the human T-HESC cell line together with the induction of interferon-β (IFN-β) and of IFN-stimulated genes. Notably, in vitro decidualization of T-HESC with cyclic AMP and progesterone upregulated the cell surface expression of the ZIKV entry co-receptor AXL and boosted ZIKV replication by ca. 100-fold. Thus, endometrial stromal cells, particularly if decidualized, likely represent a crucial cell target of ZIKV reaching them, either via the uterine vasculature in the viremic phase of the infection or by sexual viral transmission, and a potential source of virus spreading to placental trophoblasts during pregnancy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Anti-Calreticulin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Triton X-100, laboratory grade