Merck
Quantitative Trait Loci (QTL)-Guided Metabolic Engineering of a Complex Trait.
ACS synthetic biology (2016-12-13)
Matthew J Maurer, Lawrence Sutardja, Dominic Pinel, Stefan Bauer, Amanda L Muehlbauer, Tyler D Ames, Jeffrey M Skerker, Adam P Arkin
ABSTRAKT
Engineering complex phenotypes for industrial and synthetic biology applications is difficult and often confounds rational design. Bioethanol production from lignocellulosic feedstocks is a complex trait that requires multiple host systems to utilize, detoxify, and metabolize a mixture of sugars and inhibitors present in plant hydrolysates. Here, we demonstrate an integrated approach to discovering and optimizing host factors that impact fitness of Saccharomyces cerevisiae during fermentation of a Miscanthus x giganteus plant hydrolysate. We first used high-resolution Quantitative Trait Loci (QTL) mapping and systematic bulk Reciprocal Hemizygosity Analysis (bRHA) to discover 17 loci that differentiate hydrolysate tolerance between an industrially related (JAY291) and a laboratory (S288C) strain. We then used this data to identify a subset of favorable allelic loci that were most amenable for strain engineering. Guided by this "genetic blueprint", and using a dual-guide Cas9-based method to efficiently perform multikilobase locus replacements, we engineered an S288C-derived strain with superior hydrolysate tolerance than JAY291. Our methods should be generalizable to engineering any complex trait in S. cerevisiae, as well as other organisms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Antifoam 204, aqueous emulsion for bacterial and mammalian systems
Sigma-Aldrich
Glass beads, acid-washed, 425-600 μm (30-40 U.S. sieve)
Sigma-Aldrich
L-Canavanine, ≥98% (TLC), powder, from Canavalia ensiformis