- Effect of electron spectral diffusion on static dynamic nuclear polarization at 7 Tesla.
Effect of electron spectral diffusion on static dynamic nuclear polarization at 7 Tesla.
Physical chemistry chemical physics : PCCP (2017-01-18)
Alisa Leavesley, Daphna Shimon, Ting Ann Siaw, Akiva Feintuch, Daniella Goldfarb, Shimon Vega, Ilia Kaminker, Songi Han
PMID28094364
ABSTRAKT
Here, we present an integrated experimental and theoretical study of
MATERIAŁY