Merck
An in vitro skin sensitization assay termed EpiSensA for broad sets of chemicals including lipophilic chemicals and pre/pro-haptens.
Toxicology in vitro : an international journal published in association with BIBRA (2016-12-15)
Kazutoshi Saito, Osamu Takenouchi, Yuko Nukada, Masaaki Miyazawa, Hitoshi Sakaguchi
ABSTRAKT
To evaluate chemicals (e.g. lipophilic chemicals, pre/pro-haptens) that are difficult to correctly evaluate using in vitro skin sensitization tests (e.g. DPRA, KeratinoSens or h-CLAT), we developed a novel in vitro test termed "Epidermal Sensitization Assay: EpiSensA" that uses reconstructed human epidermis. This assay is based on the induction of multiple marker genes (ATF3, IL-8, DNAJB4 and GCLM) related to two keratinocyte responses (inflammatory or cytoprotective) in the induction of skin sensitization. Here, we first confirmed the mechanistic relevance of these marker genes by focusing on key molecules that regulate keratinocyte responses in vivo (P2X
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
DL-Glyceraldehyde, ≥90% (GC)
Supelco
Methyldibromoglutaronitrile, analytical standard
Sigma-Aldrich
KN-62, ≥95%, powder