Merck
  • Enhanced toxicity to the cyanobacterium Microcystis aeruginosa by low-dosage repeated exposure to the allelochemical N-phenyl-1-naphthylamine.
Enhanced toxicity to the cyanobacterium Microcystis aeruginosa by low-dosage repeated exposure to the allelochemical N-phenyl-1-naphthylamine.
Chemosphere (2017-02-19)
Y N Gao, F J Ge, L P Zhang, Y He, Z Y Lu, Y Y Zhang, B Y Liu, Q H Zhou, Z B Wu
ABSTRAKT
It has been puzzling whether and how a plant could exert a strong allelopathic inhibition to the target organisms by releasing low concentrations of allelochemicals. Plant allelochemicals have been proposed to be released continuously, however, direct evidence from specific allelochemicals is urgently required. In the present study, the toxicity of allelochemical N-phenyl-1-naphthylamine (NPN) towards the cyanobacterium Microcystis aeruginosa by two different exposure patterns was compared. One was low-dosage repeated exposure (LRE), in which 50  μg L
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-Phenyl-1-naphthylamine, reagent grade, 98%