Merck
Band-edge oscillator strength of colloidal CdSe/CdS dot-in-rods: comparison of absorption and time-resolved fluorescence spectroscopy.
Nanoscale (2017-03-23)
I Angeloni, W Raja, A Polovitsyn, F De Donato, R Proietti Zaccaria, I Moreels
ABSTRAKT
We studied the oscillator strength f
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trioctylphosphine, 97%
Sigma-Aldrich
Cadmium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Cadmium oxide, powder, 99.5% trace metals basis
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%