Merck
  • Synthesis of sandwich-like molybdenum sulfide/mesoporous organosilica nanosheets for photo-thermal conversion and stimuli-responsive drug release.
Synthesis of sandwich-like molybdenum sulfide/mesoporous organosilica nanosheets for photo-thermal conversion and stimuli-responsive drug release.
Journal of colloid and interface science (2017-02-27)
Xiaodan Su, Jin Wang, Junjie Zhang, Lihui Yuwen, Qi Zhang, Meng Dang, Jun Tao, Xiaobo Ma, Shouju Wang, Zhaogang Teng
ABSTRAKT
Sandwich-like molybdenum sulfide/mesoporous organosilica nanosheets (denoted as MoS
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,2-Bis(triethoxysilyl)ethane, 96%