Merck
Cloning and characterization of a novel intracellular protein p48.2 that negatively regulates cell cycle progression.
The international journal of biochemistry & cell biology (2009-05-12)
Fan Yang, Yu-Ping Xu, Jian Li, Su-Su Duan, Ying-Jie Fu, Yun Zhang, Yan Zhao, Wen-Tao Qiao, Qi-Min Chen, Yun-Qi Geng, Chang-Yan Che, Ying-Li Cao, Ying Wang, Lei Zhang, Lin Long, Jie He, Quan-Cai Cui, Shu-Chang Chen, Shu-Hui Wang, Li Liu
ABSTRAKT
Neurofibromatosis type 1 (NF1) microdeletion is a large genomic deletion that embraces at least 11 continuous genes at human chromosome 17q11.2. To date, most of these genes' functions still remain undefined. In this study, we report an unknown cytokine receptor like molecule (p48.2) that is frequently deleted in patients with type-1 and type-2 NF1 microdeletions in the neurofibromin locus. The cloned gene has 1317 base pair long that encodes a 438aa intracellular protein. The gene was subsequently named p48.2 based on its predicted molecular weight. A typical fibronectin type III (FNIII) domain was identified in p48.2 between Arg(176) and Pro(261) in which a palindromic Arg-Gly-Asp (RGD) repeat plus a putative Trp-Ser-X-Trp-Ser (WSXWS) motif were found at the domain's C-terminus. p48.2 mRNAs were abundant in many tumor cell lines and normal human tissues and up-regulated in some freshly isolated lung cancer and leukemia cells. Interestingly, over-expression of p48.2 in human embryo kidney 293T cells could significantly cause G0/G1 arrest and prevented S phase entry. In contrast, repressing endogenous p48.2 gene expression by specific siRNA markedly reduced G0/G1 population. Importantly, over-expression of p48.2 could significantly up-regulate rather than down-regulate cyclin D1 and cyclin D3 expressions. We further showed that the induction of cyclin D1 expression was directly due to the activation of signal transducers and activators of transcription 3 (STAT3), but was independent of RAS/mitogen-activated protein kinase (RAS/MAPK) signaling pathway. Thus, p48.2 may represent a novel type of intracellular protein functioning as a negative regulator at the G0/G1 phase.