Merck
  • Achieving a balance between small singlet-triplet energy splitting and high fluorescence radiative rate in a quinoxaline-based orange-red thermally activated delayed fluorescence emitter.
Achieving a balance between small singlet-triplet energy splitting and high fluorescence radiative rate in a quinoxaline-based orange-red thermally activated delayed fluorescence emitter.
Chemical communications (Cambridge, England) (2016-08-20)
Ling Yu, Zhongbin Wu, Guohua Xie, Cheng Zhong, Zece Zhu, Hengjiang Cong, Dongge Ma, Chuluo Yang
ABSTRAKT
A new orange-red thermally activated delayed fluorescence (TADF) emitter is designed and synthesized by incorporating a fluorine-substituted quinoxaline as an electron-acceptor and a phenoxazine as an electron-donor. The rational molecular design enables small singlet-triplet energy splitting (ΔEST) and high fluorescence radiative rate (k) for long-wavelength TADF emitters. The organic light emitting diodes (OLEDs) employing the new TADF emitter achieve maximum external quantum efficiencies (EQEs) of 13.9% and 9.0% for the vacuum- and solution-processed OLEDs, respectively.