- Nearly 100% triplet harvesting in conventional fluorescent dopant-based organic light-emitting devices through energy transfer from exciplex.
Nearly 100% triplet harvesting in conventional fluorescent dopant-based organic light-emitting devices through energy transfer from exciplex.
Advanced materials (Deerfield Beach, Fla.) (2015-02-14)
Xiao-Ke Liu, Zhan Chen, Cai-Jun Zheng, Miao Chen, Wei Liu, Xiao-Hong Zhang, Chun-Sing Lee
PMID25676085
ABSTRAKT
Nearly 100% triplet harvesting in conventional fluorophor-based organic light-emitting devices is realized through energy transfer from exciplex. The best C545T-doped device using the exciplex host exhibits a maximum current efficiency of 44.0 cd A(-1) , a maximum power efficiency of 46.1 lm W(-1) , and a maximum external quantum efficiency of 14.5%.