Merck
NPM1 directs PIDDosome-dependent caspase-2 activation in the nucleolus.
The Journal of cell biology (2017-04-23)
Kiyohiro Ando, Melissa J Parsons, Richa B Shah, Chloé I Charendoff, Sheré L Paris, Peter H Liu, Sara R Fassio, Brittany A Rohrman, Ruth Thompson, Andrew Oberst, Samuel Sidi, Lisa Bouchier-Hayes
ABSTRAKT
The PIDDosome (PIDD-RAIDD-caspase-2 complex) is considered to be the primary signaling platform for caspase-2 activation in response to genotoxic stress. Yet studies of PIDD-deficient mice show that caspase-2 activation can proceed in the absence of PIDD. Here we show that DNA damage induces the assembly of at least two distinct activation platforms for caspase-2: a cytoplasmic platform that is RAIDD dependent but PIDD independent, and a nucleolar platform that requires both PIDD and RAIDD. Furthermore, the nucleolar phosphoprotein nucleophosmin (NPM1) acts as a scaffold for PIDD and is essential for PIDDosome assembly in the nucleolus after DNA damage. Inhibition of NPM1 impairs caspase-2 processing, apoptosis, and caspase-2-dependent inhibition of cell growth, demonstrating that the NPM1-dependent nucleolar PIDDosome is a key initiator of the caspase-2 activation cascade. Thus we have identified the nucleolus as a novel site for caspase-2 activation and function.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
MISSION® esiRNA, targeting human NPM1