Merck
  • Selective solid phase extraction of chloroacetamide herbicides from environmental water samples by amphiphilic magnetic molecularly imprinted polymers.
Selective solid phase extraction of chloroacetamide herbicides from environmental water samples by amphiphilic magnetic molecularly imprinted polymers.
Talanta (2017-05-16)
Wenhua Ji, Rihan Sun, Wenjuan Duan, Xiao Wang, Tao Wang, Yan Mu, Lanping Guo
ABSTRAKT
In this study, a novel amphiphilic magnetic molecularly imprinted polymers (MMIPs) have been prepared by using Fe
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Acetochlor, PESTANAL®, analytical standard
Supelco
Propisochlor, PESTANAL®, analytical standard
Supelco
Pretilachlor, PESTANAL®, analytical standard