- Angiotensin II receptor blockers induce autophagy in prostate cancer cells.
Angiotensin II receptor blockers induce autophagy in prostate cancer cells.
Oncology letters (2017-05-23)
Yunseo Woo, Yu-Jin Jung
PMID28529582
ABSTRAKT
Angiotensin II receptor blockers (ARBs) are anti-hypertensive drugs that competitively inhibit the binding of angiotensin II to its receptor, resulting in blood vessel dilation and the reduction of blood pressure. These antagonists are also known as sartans, and are a group of pharmaceuticals that possess tetrazole or imidazole groups. In the present study, the anticancer and antimetastatic effects of the ARBs fimasartan, losartan, eprosartan and valsartan on the human prostate cancer PC-3, DU-145 and LNCap-LN3 cell lines were investigated