Merck
Novel cellulose-based halochromic test strips for naked-eye detection of alkaline vapors and analytes.
Talanta (2017-05-16)
Hussein Abou-Yousef, Tawfik A Khattab, Yehia A Youssef, Naser Al-Balakocy, Samir Kamel
ABSTRAKT
A simple, portable and highly sensitive naked-eye test strip is successfully prepared for optical detection of gaseous and aqueous alkaline analytes. Novel pH-sensory tricyanofuran-hydrazone (TCFH) disperse colorant containing a hydrazone recognition functional moiety is successfully synthesized via azo-coupling reaction between active methyl-containing tricyanofuran (TCF) heterocycle and diazonium salt of 4-aminobenzaldehyde followed by Knoevenagel condensation with malononitrile. UV-vis absorption spectra display solvatochromism and reversible color changes of the TCFH solution in dimethyl sulfoxide in response to pH variations. We investigate the preparation of hydrophobic cellulose/polyethylene terephthalate composites characterized by their high affinity for disperse dyes. Composite films made from CA, Cell/CA, PET/CA, and Cell/PET-CA are produced via solvent-casting procedure using 10-30% modified cellulose or modified polyethylene terephthalate. The mechanical properties and morphologies of these composite films are investigated. The prepared pH-sensory hydrazone-based disperse dye is then applied to dye the produced cellulose-based composite films employing the high temperature pressure dyeing procedure. The produced halochromic PET-CA-TCFH test strip provide an instant visible signal from orange to purple upon exposure to alkaline conditions as proved by the coloration measurements. The sensor strip exhibits high sensitivity and quick detection toward ammonia in both of aqueous and vapor phases by naked-eye observations at room temperature and atmospheric pressure.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1,3-Propylene sulfite, 99%