- The interactive role of CB1 receptors and L-type calcium channels in hippocampal long-term potentiation in rats.
The interactive role of CB1 receptors and L-type calcium channels in hippocampal long-term potentiation in rats.
Brain research bulletin (2017-04-27)
Hamidreza Komaki, Fargol Saadat, Siamak Shahidi, Abdolrahman Sarihi, Parisa Hasanein, Alireza Komaki
PMID28442324
ABSTRAKT
Long-term potentiation (LTP) of synaptic responses is a widely researched model of synaptic plasticity that occurs during learning and memory. The cannabinoid system is an endogenous system that modulate this kind of synaptic plasticity. In addition, voltage dependent calcium channels is essential for induction of LTP at some synapses in the hippocampus. However, there is currently debate over the interaction between L-type calcium channels and cannabinoid system on the synaptic plasticity. In this study, we examined the effects of an acute administration of the cannabinoid antagonist AM251 following a chronic administration of the Ca
MATERIAŁY